missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vandetanib-d5 (Major), TRC
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
Specifications
Specifications
| Chemical Name or Material | Vandetanib-d5 |
| Molecular Formula | C22 D5 H19 Br F N4 O2 |
| Quantity | 250 mg |
| Synonym | N-(4-bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl-2,3,4,5,6-d5)methoxy)quinazolin-4-amine |
| InChI Formula | InChI=1S/C22H24BrFN4O2/c1-28-7-5-14(6-8-28)12-30-21-11-19-16(10-20(21)29-2)22(26-13-25-19)27-18-4-3-15(23)9-17(18)24/h3-4,9-11,13-14H,5-8,12H2,1-2H3,(H,25,26,27)/i5D,6D,7D,8D,14D |
| SMILES | [2H]C1C([2H])C([2H])(COc2cc3ncnc(Nc4ccc(Br)cc4F)c3cc2OC)C([2H])C([2H])N1C |
| IUPAC Name | N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(2,3,4,5,6-pentadeuterio-1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine |
| Molecular Weight (g/mol) | 480.385 |
| Formula Weight | 479.138 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction