missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Standard: CH3CO2- @ 1000 μg/mL in H2O, VHG™
Description
Acetate Standard: CH3CO2- @ 1000 μg/mL in H2O
Specifications
Specifications
| Chemical Name or Material | Acetate |
| CAS | 71-50-1 |
| Methods | Ion Chromatography |
| Analyte or Component Names | Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H3 O2 |
| Synonym | (C2 H3 O)2- |
| InChI Formula | InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/p-1 |
| SMILES | CC(=O)[O-] |
| IUPAC Name | acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?