missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| CAS | 87269-87-2 |
| Analyte or Component Names | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| InChI Formula | InChI=1S/C15H19NO2.ClH/c17-15(18-10-11-5-2-1-3-6-11)14-9-12-7-4-8-13(12)16-14;/h1-3,5-6,12-14,16H,4,7-10H2;1H/t12-,13-,14-;/m0./s1 |
| SMILES | Cl.O=C(OCc1ccccc1)[C@@H]2C[C@@H]3CCC[C@@H]3N2 |
| IUPAC Name | benzyl (2S,3aS,6aS)-1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrole-2-carboxylate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction