missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride (Risperidone Piperidopyrimidinone Intermediate Hydrochloride), Mikromol™
Description
3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride (Risperidone Piperidopyrimidinone Intermediate Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride |
| CAS | 93076-03-0 |
| Analyte or Component Names | 3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 Cl N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Risperidone Imp. L (EP), 3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride, Risperidone Imp. L (EP) as Hydrochloride, Risperidone Piperidopyrimidinone Intermediate Hydrochloride, Risperidone Impurity L as Hydrochloride |
| InChI Formula | InChI=1S/C11H15ClN2O.ClH/c1-8-9(5-6-12)11(15)14-7-3-2-4-10(14)13-8;/h2-7H2,1H3;1H |
| SMILES | Cl.CC1=C(CCCl)C(=O)N2CCCCC2=N1 |
| IUPAC Name | 3-(2-chloroethyl)-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?