missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human c-Myc Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB3696-SP
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| c-Myc | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, Flow Cytometry 2.5 μg/10^6 cells, Immunohistochemistry 8-25 μg/mL, Immunoprecipitation, Immunocytochemistry 8-25 μg/mL, CyTOF-ready, Knockout Validated | |
| P01106 | |
| MYC | |
| C-terminal region peptide of human c-Myc Ala408-Ala439 Accession # P01106 | |
| 25 μg | |
| Primary | |
| Detects endogenous human c-Myc and c-Myc tagged proteins in Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF, Immunoassay | |
| 90000000000 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, cMyc, c-Myc, MYC, myc proto-oncogene protein, Myc2, MYCC, myc-related translation/localization regulatory factor, Niard, Nird, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4609 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction