missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
610.00€
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Form | Purified |
Description
2B4/CD244/SLAMF4 Polyclonal antibody specifically detects 2B4/CD244/SLAMF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| 2B4/CD244/SLAMF4 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human SLAMF4. The immunogen is located within the last 50 amino acids of SLAMF4. Amino Acid Squence: PKAQNPARLSRKELEN | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| PerCP | |
| Rabbit | |
| Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| PBS | |
| 51744 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title