missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 2D Bar-coded Cryogenic Vial Storage Box for 81 Vials

Product Code. 15525729 Shop All Corning Products
Change view
Click to view available options
Holds:
81 (2 mL Vials)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15525729 81 (2 mL Vials) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15525729 Supplier Corning™ Supplier No. 8673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For storing cryogenic vials. Corning™ 2D Bar-coded Cryogenic Vial Storage Box is made of polycarbonate.

  • Reusable white polycarbonate box holds 81 cryogenic vials in a 9 x 9 array
  • Compatible with both internal and external threaded vials
TRUSTED_SUSTAINABILITY

Specifications

Type Cryo Storage Box
Holds 81 (2 mL Vials)
Tube Array 9 x 9
Weight (Metric) 12 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.