missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-Methyl-4-(benzyloxy)-2-butenoic Acid Methyl Ester, TRC

Product Code. 30475611
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475611 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475611 Supplier Toronto Research Chemicals Supplier No. TRCM28897510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-3-Methyl-4-(benzyloxy)-2-butenoic Acid Methyl Ester
CAS 310887-98-0
Molecular Formula C13H16O3
Quantity 10 mg
Synonym (2E)-3-Methyl-4-(phenylmethoxy)-2-butenoic Acid Methyl Ester
InChI Formula InChI=1S/C13H16O3/c1-11(8-13(14)15-2)9-16-10-12-6-4-3-5-7-12/h3-8H,9-10H2,1-2H3/b11-8+
SMILES O=C(OC)/C=C(C)/COCC1=CC=CC=C1
IUPAC Name methyl (E)-4-(benzyloxy)-3-methylbut-2-enoate
Molecular Weight (g/mol) 220.26
Formula Weight 220.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.