Learn More
2H-Furo[2,3-c]pyran-2-one, TRC
Product Code. 30012763
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012763 1 mg 1mg
30006289 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30012763 Supplier Toronto Research Chemicals Supplier No. TRCF8648001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 857054-03-6
Molecular Formula C7H4O3
Molecular Weight (g/mol) 136.1
Synonym KAR2, Karrikin 2;
SMILES C1=COC=C2C1=CC(=O)O2
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C7H4O3/c8-7-3-5-1-2-9-4-6(5)10-7/h1-4H
Percent Purity >95%
Chemical Name or Material 2H-Furo[2,3-c]pyran-2-one
Grade HPLC