missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-2-Methylbutane-1,2,3,4-tetraol, TRC

Product Code. 30133138
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133138 100 mg 100mg
30012449 10 mg 10mg
30010369 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133138 Supplier Toronto Research Chemicals Supplier No. TRCM295130100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 310887-92-4
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.15
SMILES OC[C@](O)(C)[C@@H](O)CO

Specifications

Quantity 100 mg
Chemical Name or Material (2R,3R)-2-Methylbutane-1,2,3,4-tetraol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.