missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (2R,4R)-APDC
GREENER_CHOICE

Product Code. 18035332
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035332 10 mg 10mg
30001421 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18035332 Supplier Tocris Bioscience™ Supplier No. 1208/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective group II agonist

A highly selective and relatively potent group II metabotropic glutamate receptor agonist. EC50 values are 0.4, 0.4, > 100, > 100, > 300 and > 300 μM for human mGlu2, mGlu3, mGlu1, mGlu5, mGlu4 and mGlu7 receptors respectively. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169209-63-6
Molecular Formula C6H10N2O4

Specifications

Quantity 10 mg
Formula Weight 174.16
Purity >98%
Molecular Weight (g/mol) 174.156
Chemical Name or Material (2R, 4R)-APDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.