Learn More
(2R,6S)-rel-2,6-Dimethyl-4-(2-methyl-3-phenylpropyl)morpholine Hydrochloride, TRC
Product Code. 30008329
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008329 250 mg 250mg
30007402 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008329 Supplier Toronto Research Chemicals Supplier No. TRCD474815250MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 922734-43-8
Molecular Formula C16H26ClNO
Molecular Weight (g/mol) 283.84
Synonym (±)-2,6-cis-Dimethyl-4-(2-methyl-3-phenylpropyl)-morpholine Hydrochloride, Amorolfine EP Impurity C
SMILES Cl.CC(CN1C[C@@H](C)O[C@@H](C)C1)Cc2ccccc2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C16H25NO.ClH/c1-13(9-16-7-5-4-6-8-16)10-17-11-14(2)18-15(3)12-17;/h4-8,13-15H,9-12H2,1-3H3;1H/t13?,14-,15+;
Formula Weight 283.1703
Chemical Name or Material (2R,6S)-rel-2,6-Dimethyl-4-(2-methyl-3-phenylpropyl)morpholine Hydrochloride