missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride, Mikromol™

Product Code. 18051210
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18051210 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18051210 Supplier Mikromol Supplier No. MM0022.120025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride
CAS 1794-55-4
Analyte or Component Names (2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C26 H36 N2 O4 . Cl H
Certifications/Compliance ISO 9001
Synonym Verapamil Hydrochloride Imp. I (EP), Verapamil Imp. I (EP), Verapamil USP Related Compound B, Verapamil USP RC B, (2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride, Verapamil Hydrochloride Imp. I (EP) as Hydrochloride, Verapamil Related Compound B as Hydrochloride, Verapamil Hydrochloride Impurity I as Hydrochloride, Verapamil Impurity I as Hydrochloride
InChI Formula InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-5)25(17-21)32-7)13-15-28(3)14-12-20-8-10-22(29-4)24(16-20)31-6;/h8-11,16-17,19H,12-15H2,1-7H3;1H
SMILES Cl.COc1ccc(CCN(C)CCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC
IUPAC Name 2-(3,4-dimethoxyphenyl)-2-[2-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]ethyl]-3-methylbutanenitrile;hydrochloride
Molecular Weight (g/mol) 477.04
Quantity 25 mg
Formula Weight 476.2442 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.