missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, Mikromol™

Product Code. 30049980
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049980 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049980 Supplier Mikromol Supplier No. MM0613.04
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide
CAS 1037184-07-8
Analyte or Component Names (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide
Shipping Condition Room Temperature
Molecular Formula C14 H19 Cl2 N O
Certifications/Compliance ISO 9001
Synonym Bupivacaine Hydrochloride Imp. D (EP), Bupivacaine Imp. D (EP), (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, Bupivacaine Hydrochloride Impurity D, Bupivacaine Impurity D
InChI Formula InChI=1S/C14H19Cl2NO/c1-10-6-5-7-11(2)13(10)17-14(18)12(16)8-3-4-9-15/h5-7,12H,3-4,8-9H2,1-2H3,(H,17,18)
SMILES Cc1cccc(C)c1NC(=O)C(Cl)CCCCCl
IUPAC Name 2,6-dichloro-N-(2,6-dimethylphenyl)hexanamide
Molecular Weight (g/mol) 288.21
Quantity 50 mg
Formula Weight 287.0844 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.