missing translation for 'onlineSavingsMsg'
Learn More

(2S)-1-[(2S)-2-Aminopropanoyl]pyrrolidine-2-carboxylic Acid (L-Alanyl-L-proline), Mikromol™

Product Code. 30050918
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050918 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050918 Supplier Mikromol Supplier No. MM0010.270025
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-1-[(2S)-2-Aminopropanoyl]pyrrolidine-2-carboxylic Acid (L-Alanyl-L-proline)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Alanyl-L-proline
CAS 13485-59-1
Analyte or Component Names L-Alanyl-L-proline
Shipping Condition Room Temperature
Molecular Formula C8 H14 N2 O3
Synonym L-Proline, L-alanyl-, L-Proline, 1-L-alanyl-, Proline, 1-L-alanyl- (7CI), Proline, 1-L-alanyl-, L- (8CI), L-Alanyl-L-proline, 101: PN: US20070066537 PAGE: 17 claimed protein, 12: PN: WO2014170713 SEQID: 112 claimed protein, 150: PN: EP2161028 PAGE: 10 claimed protein, 443: PN: WO2005081628 SEQID: 445 claimed protein, 76: PN: WO2011146121 PAGE: 115 claimed sequence, Alanylproline, L-α-Alanyl-L-proline, N-Alanylproline, NSC 97932
InChI Formula InChI=1S/C8H14N2O3/c1-5(9)7(11)10-4-2-3-6(10)8(12)13/h5-6H,2-4,9H2,1H3,(H,12,13)/t5-,6-/m0/s1
SMILES C[C@H](N)C(=O)N1CCC[C@H]1C(=O)O
IUPAC Name (2S)-1-[(2S)-2-aminopropanoyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 186.21
Quantity 25 mg
Formula Weight 186.1004 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.