missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-[[(4S)-4-[[4-[2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]-4-carboxybutanoyl]amino]pentanedioic Acid, Mikromol
Description
(2S)-2-[[(4S)-4-[[4-[2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]-4-carboxybutanoyl]amino]pentanedioic Acid, Mikromol
Specifications
Specifications
| Chemical Name or Material | (2S)-2-(((4S)-4-((4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo(2,3-d)pyrimidin-5-yl)ethyl)benzoyl)amino)-4-carboxybutanoyl)amino)pentanedioic Acid |
| CAS | 144051-68-3 |
| Analyte or Component Names | (2S)-2-(((4S)-4-((4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo(2,3-d)pyrimidin-5-yl)ethyl)benzoyl)amino)-4-carboxybutanoyl)amino)pentanedioic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H28 N6 O9 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-γ-glutamyl-, L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-γ-glutamyl- (9CI), L-Glutamic acid, N-[N |
| InChI Formula | InChI=1S/C25H28N6O9/c26-25-30-20-19(22(36)31-25)14(11-27-20)6-3-12-1-4-13(5-2-12)21(35)29-16(24(39)40)7-9-17(32)28-15(23(37)38)8-10-18(33)34/h1-2,4-5,11,15-16H,3,6-10H2,(H,28,32)(H,29,35)(H,33,34)(H,37,38)(H,39,40)(H4,26,27,30,31,36)/t15-,16-/m0/s1 |
| Solution Type | Neat |
| SMILES | NC1=NC(=O)c2c(CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)O)c[nH]c2N1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?