Learn More
(2S,3R)-3-Hydroxynorleucine, TRC
Product Code. 30400459
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400459 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30400459 Supplier Toronto Research Chemicals Supplier No. TRCH94976010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,3R)-3-Hydroxynorleucine
CAS 10148-69-3
Molecular Formula C6H13NO3
Quantity 10 mg
InChI Formula InChI=1S/C6H13NO3/c1-2-3-4(8)5(7)6(9)10/h4-5,8H,2-3,7H2,1H3,(H,9,10)/t4-,5+/m1/s1
SMILES N[C@H](C(O)=O)[C@H](O)CCC
IUPAC Name (2S,3R)-2-amino-3-hydroxyhexanoic acid
Molecular Weight (g/mol) 147.17
Formula Weight 147.0895