missing translation for 'onlineSavingsMsg'
Learn More

3-[1-(Methylamino)ethyl]phenyl N-ethyl-N-methylcarbamate, TRC

Product Code. 30133089
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133089 10 mg 10mg
30133523 100 mg 100mg
18082940 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133089 Supplier Toronto Research Chemicals Supplier No. TRCM13800510MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948051-93-2
Molecular Formula C13 H20 N2 O2
Molecular Weight (g/mol) 236.31
Synonym Carbamic acid, N-ethyl-N-methyl-, 3-[1-(methylamino)ethyl]phenyl ester, 3-[1-(Methylamino)ethyl]phenyl N-ethyl-N-methylcarbamate, Rivastigmine Tartrate Imp E (EP) as Racemate
IUPAC Name [3-[1-(methylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate
SMILES CCN(C)C(=O)Oc1cccc(c1)C(C)NC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H20N2O2/c1-5-15(4)13(16)17-12-8-6-7-11(9-12)10(2)14-3/h6-10,14H,5H2,1-4H3
Formula Weight 236.1525
Chemical Name or Material 3-[1-(Methylamino)ethyl]phenyl N-ethyl-N-methylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.