missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-yl)benzoic acid, 97%

Product Code. 15493717
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493717 250 mg 250mg
11466480 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493717 Supplier Thermo Scientific Alfa Aesar Supplier No. H27581.MD
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73096-39-6 | C8H6N4O2 | 190.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73096-39-6
Molecular Formula C8H6N4O2
Molecular Weight (g/mol) 190.162
MDL Number MFCD03821131
InChI Key MIERACSHCALJOM-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl benzoic acid, 3-2h-tetrazol-5-yl benzoic acid, 3-1h-tetrazol-5-yl-benzoic acid, 3-2h-1,2,3,4-tetrazol-5-yl benzoic acid, benzoic acid, 3-1h-tetrazol-5-yl, 3-1h-1,2,3,4-tetrazol-5-yl benzoic acid, 3-2h-tetrazol-5-yl-benzoic acid, 3-1h-1,2,3,4-tetraazol-5-yl benzoic acid, 3-1h-tetrazol-5-yl benzoicacid, 3-1h-tetrazole-5-yl benzoic acid
PubChem CID 333942
IUPAC Name 3-(2H-tetrazol-5-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=NNN=N2

Specifications

Melting Point 284°C to 286°C
Quantity 250 mg
Formula Weight 190.16
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-yl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.