missing translation for 'onlineSavingsMsg'
Learn More

3-[(1S)-1-(Dimethylamino)ethyl]phenyl Diethylcarbamate, Mikromol™

Product Code. 30049216
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049216 Supplier Mikromol Supplier No. MM1497.160100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[(1S)-1-(Dimethylamino)ethyl]phenyl Diethylcarbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Diethylcarbamate
CAS 1230021-34-7
Analyte or Component Names 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Diethylcarbamate
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H24 N2 O2
Certifications/Compliance ISO 9001
Synonym Carbamic acid, N,N-diethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester
InChI Formula InChI=1S/C15H24N2O2/c1-6-17(7-2)15(18)19-14-10-8-9-13(11-14)12(3)16(4)5/h8-12H,6-7H2,1-5H3/t12-/m0/s1
SMILES CCN(CC)C(=O)Oc1cccc(c1)[C@H](C)N(C)C
IUPAC Name [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N,N-diethylcarbamate
Molecular Weight (g/mol) 264.3633
Quantity 100 mg
Formula Weight 264.1838 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.