missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[2-[4-(6-Hydroxy-1,2-benzoxazol-3-yl)-1-oxidopiperidin-1-ium-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one (6-Desfluoro-6-hydroxyrisperidone N-Oxide), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-[2-[4-(6-Hydroxy-1,2-benzoxazol-3-yl)-1-oxidopiperidin-1-ium-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one (6-Desfluoro-6-hydroxyrisperidone N-Oxide)
Specifications
Specifications
| Chemical Name or Material | 6-Desfluoro-6-hydroxyrisperidone N-Oxide |
| CAS | 832747-58-7 |
| Analyte or Component Names | 6-Desfluoro-6-hydroxyrisperidone N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 N4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4H-Pyrido[1,2-a]pyrimidin-4-one, 6,7,8,9-tetrahydro-3-[2-[4-(6-hydroxy-1,2-benzisoxazol-3-yl)-1-oxido-1-piperidinyl]ethyl]-2-methyl-, 3-[2-[4-(6-Hydroxy-1,2-benzisoxazol-3-yl)-1-oxidopiperidin-1-yl]ethyl]-2-methyl-6,7,8,9- tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, 6,7,8,9-Tetrahydro-3-[2-[4-(6-hydroxy-1,2-benzisoxazol-3-yl)-1-oxido-1-piperidinyl]ethyl]-2-methyl- 4H-pyrido[1,2-a]pyrimidin-4-one |
| InChI Formula | InChI=1S/C23H28N4O4/c1-15-18(23(29)26-10-3-2-4-21(26)24-15)9-13-27(30)11-7-16(8-12-27)22-19-6-5-17(28)14-20(19)31-25-22/h5-6,14,16,28H,2-4,7-13H2,1H3 |
| SMILES | CC1=C(CC[N+]2([O-])CCC(CC2)c3noc4cc(O)ccc34)C(=O)N5CCCCC5=N1 |
| IUPAC Name | 3-[2-[4-(6-hydroxy-1,2-benzoxazol-3-yl)-1-oxidopiperidin-1-ium-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction