missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-thiazol-4-yl)-benzaldehyde, 97%, Thermo Scientific™

Product Code. 10316354
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10316354 5 g 5g
10769793 250 mg 250mg
10168172 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316354 Supplier Thermo Scientific Maybridge Supplier No. CC19404DE
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-05-2
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.26
MDL Number MFCD02681946
InChI Key FQQPVEKQPGDYPD-UHFFFAOYSA-N
PubChem CID 7127782
IUPAC Name 3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
SMILES CC1=NC(=CS1)C1=CC(C=O)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 3-2-methyl-thiazol-4-yl-benzaldehyde, 3-2-methyl-1,3-thiazol-4-yl benzaldehyde, 3-2-methylthiazol-4-yl benzaldehyde, 4-3-formylphenyl-2-methyl-1,3-thiazole, 3-2-methyl-4-thiazolyl benzaldehyde, benzaldehyde,3-2-methyl-4-thiazolyl, benzaldehyde, 3-2-methyl-4-thiazolyl
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-thiazol-4-yl)-benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.