missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%, Thermo Scientific™

Product Code. 15440636
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440636 1 g 1g
15450636 5 g 5g
15460636 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440636 Supplier Thermo Scientific Alfa Aesar Supplier No. H53388.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 564-10-3
Molecular Formula C4H2F6O2
Molecular Weight (g/mol) 196.048
MDL Number MFCD00464165
InChI Key RAEAYTICAPHWJW-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
PubChem CID 385606
IUPAC Name 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
SMILES C(C(=O)O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 46°C to 53°C
Boiling Point 125°C
Linear Formula (CF3)2CHCO2H
Quantity 1 g
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.