missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluorolactic acid, 96%, Thermo Scientific Chemicals

Product Code. 11332037
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332037 1 g 1g
11342037 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332037 Supplier Thermo Scientific Alfa Aesar Supplier No. L07742.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluorolactic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, strong bases and water. Protect from humidity and water. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 684-07-1
Molecular Formula C3H3F3O3
Molecular Weight (g/mol) 144.049
MDL Number MFCD00051802
InChI Key BVKGUTLIPHZYCX-UHFFFAOYSA-N
Synonym 3,3,3-trifluorolactic acid, trifluorolactic acid, 2-hydroxy-3,3,3-trifluoropropanoic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy, 2-hydroxy-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-hydroxypropionic acid, acmc-20epmv, acmc-20dpo7, propanoic acid,3,3,3-trifluoro-2-hydroxy-, 2r, 3,3,3-trifluoro-dl-lactic acid
PubChem CID 244896
IUPAC Name 3,3,3-trifluoro-2-hydroxypropanoic acid
SMILES C(C(=O)O)(C(F)(F)F)O

Specifications

Melting Point 66°C to 71°C
Boiling Point 157°C (123 mmHg)
Quantity 1 g
UN Number UN3261
Beilstein 1907003
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 144.05
Percent Purity 96%
Chemical Name or Material 3,3,3-Trifluorolactic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.