missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4',5-Pentachlorobiphenyl, TRC

Product Code. 30136054
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136054 50 mg 50mg
30132585 25 mg 25mg
17997369 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136054 Supplier Toronto Research Chemicals Supplier No. TRCP27449350MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57465-28-8
Molecular Formula C12 H5 Cl5
Molecular Weight (g/mol) 326.43
Synonym 1,1'-Biphenyl, 3,3',4,4',5-pentachloro-, 3,3',4,4',5-Pentachloro-1,1'-biphenyl, 3,3',4,4',5-Pentachlorobiphenyl, 3,3',4,4',5'-Pentachlorobiphenyl, 3,4,3',4',5'-Pentachlorobiphenyl, 3,4,5,3',4'-Pentachlorobiphenyl, CB 126, PCB 126, PnCP 176, Polychlorinated biphenyl-126, 3,3',4,4',5-PentaCB
IUPAC Name 1,2,3-trichloro-5-(3,4-dichlorophenyl)benzene
SMILES Clc1ccc(cc1Cl)c2cc(Cl)c(Cl)c(Cl)c2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H5Cl5/c13-8-2-1-6(3-9(8)14)7-4-10(15)12(17)11(16)5-7/h1-5H
Formula Weight 323.8834
Percent Purity >95%
Chemical Name or Material PCB No. 126
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.