Learn More
3-(3,4,5-Trimethoxyphenyl)propanoic acid, 97%, BLD Pharm™
Product Code. 30941099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30941099 100 g 100g
30930190 1 g 1g
30935550 5 g 5g
30934860 10 g 10g
30940379 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30941099 Supplier BLD Pharm Supplier No. BD14165100g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 25173-72-2
Molecular Formula C12H16O5
MDL Number MFCD00002775
Quantity 100 g
SMILES O=C(O)CCC1=CC(OC)=C(OC)C(OC)=C1
Molecular Weight (g/mol) 240.25
Percent Purity 97%
Chemical Name or Material 3-(3,4,5-Trimethoxyphenyl)propanoic acid