Learn More
3,3-Dimethoxy-2-butanone, 98%
Product Code. 10284814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10284814 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10284814 Supplier Thermo Scientific Acros Supplier No. 399720250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
CAS: 21983-72-2 | C6H12O3 | 132.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 21983-72-2
Density 0.9870g/mL
Boiling Point 145°C to 146°C
Flash Point 45°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H12O3
Linear Formula CH3C(OCH3)2C(O)CH3
MDL Number MFCD00075266
Quantity 25 g
Specific Gravity 0.987
Synonym 3,3-dimethoxy-2-butanone, 2-butanone, 3,3-dimethoxy, acmc-20aa6y, 3.3-dimethoxy-2-butanone, 3,3-dimethoxy-2-oxobutane, 3,3-dimethoxy-butan-2-one, ksc495e8l
InChI Key UFQBSPGKRRSATO-UHFFFAOYSA-N
SMILES COC(C)(OC)C(C)=O
IUPAC Name 3,3-dimethoxybutan-2-one
Molecular Weight (g/mol) 132.16
PubChem CID 140871
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 3, 3-Dimethoxy-2-butanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethoxy-2-butanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.