Learn More
3,3'-Dimethoxybenzidine, 97%
Product Code. 10269880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10269880 5 g Glass bottle 5g
10065810 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10269880 Supplier Thermo Scientific Acros Supplier No. 407890050
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 119-90-4 | C14H16N2O2 | 244.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119-90-4
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008372
InChI Key JRBJSXQPQWSCCF-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
PubChem CID 8411
ChEBI CHEBI:82321
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N
Specifications
CAS Min % 96.0
CAS Max % 100.0
Melting Point 135°C to 139°C
Color Beige-Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 2992
Formula Weight 244.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3'-Dimethoxybenzidine, pract., 97%