missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzylamine, 97%, Thermo Scientific™

Product Code. 10701894
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10701894 250 mg Amber Glass Bottle 250mg
10682885 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10701894 Supplier Thermo Scientific Maybridge Supplier No. CC59413CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 182963-94-6
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.31
MDL Number MFCD09064990
InChI Key VRMIFLYRNDVHIN-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy benzylamine, 3-3-aminomethyl phenoxy-n,n-dimethylpropan-1-amine, 3-3-dimethylamino propoxy phenyl methanamine, 3-3-aminomethyl phenoxy propyl dimethylamine, benzenemethanamine,3-3-dimethylamino propoxy, 3-3-aminomethyl phenoxy-n,n-dimethyl-propan-1-amine
PubChem CID 21465371
SMILES CN(C)CCCOC1=CC=CC(CN)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 3-[3-(aminomethyl)phenoxy]-N,N-dimethylpropan-1-amine
Formula Weight 208.3
Percent Purity 95%
Chemical Name or Material 3-[3-(Dimethylamino)propoxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.