missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Furyl)acrylic acid, 98%

Product Code. 11377926
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377926 25 g 25g
11357926 1 g 1g
11367926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377926 Supplier Thermo Scientific Alfa Aesar Supplier No. L13143.14
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 39244-10-5 | C7H6O3 | 138.12 g/mol

3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

Solubility
Sparingly soluble in water (0.56 g/L at 25°C).

Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39244-10-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00075074
InChI Key JHAPZUDWRRBZHZ-UHFFFAOYSA-N
Synonym 3-3-furyl acrylic acid, 3-furan-3-yl acrylic acid, trans-3-furanacrylic acid, e-3-furan-3-yl acrylic acid, 2-propenoic acid, 3-3-furanyl-, 2e, 2e-3-3-furyl prop-2-enoic acid, 2e-3-furan-3-yl prop-2-enoic acid, 3-3-furanyl-2-propenoic acid, 2e-3-3-furyl acrylic acid, furan-3-acrylic acid
PubChem CID 643403
IUPAC Name (E)-3-(furan-3-yl)prop-2-enoic acid
SMILES OC(=O)C=CC1=COC=C1

Specifications

Melting Point 152°C to 154°C
Quantity 25 g
Beilstein 1306450
Solubility Information Sparingly soluble in water (0.56g/L at 25°C).
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 3-(3-Furyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.