missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Iminobis(N,N-dimethylpropylamine)

Product Code. 17261222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17261222 25 mL 25mL
17271222 100 mL 100mL
17281222 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17261222 Supplier Thermo Scientific Acros Supplier No. 466340250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6711-48-4 | C10H25N3 | 187.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6711-48-4
Molecular Formula C10H25N3
Molecular Weight (g/mol) 187.33
InChI Key BXYVQNNEFZOBOZ-UHFFFAOYSA-N
IUPAC Name (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine
SMILES CN(C)CCCNCCCN(C)C

Specifications

Melting Point -78.0°C
Density 0.8410 g/mL
Boiling Point 128.0°C to 131.0°C (27.0 hPa)
Flash Point 98°C
Quantity 25 mL
Specific Gravity 0.841
Formula Weight 187.33
Chemical Name or Material 3,3 '-Iminobis(N,N-dimethylpropylamine)
compliance-icons compliance-icons
Product Identifier
  • 3,3'-Iminobis(N,N-dimethylpropylamine)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3,3'-Iminobis(N,N-dimethylpropylamine)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.