missing translation for 'onlineSavingsMsg'
Learn More

(+)-3,4,4a,5,6,10b-Hexahydro-2H-naphtho[1,2-b][1,4]oxazin-9-ol Hydrochloride, TRC

Product Code. 30135600
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135600 2.5 mg 2.5mg
30133688 25 mg 25mg
30008401 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135600 Supplier Toronto Research Chemicals Supplier No. TRCH2940752.5MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 858517-21-2
Molecular Formula C12 H15 N O2 . Cl H
Molecular Weight (g/mol) 241.71
IUPAC Name (4aR,10bR)-3,4,4a,5,6,10b-hexahydro-2H-benzo[h][1,4]benzoxazin-9-ol;hydrochloride
SMILES Cl.Oc1ccc2CC[C@H]3NCCO[C@@H]3c2c1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C12H15NO2.ClH/c14-9-3-1-8-2-4-11-12(10(8)7-9)15-6-5-13-11;/h1,3,7,11-14H,2,4-6H2;1H/t11-,12-;/m1./s1
Formula Weight 241.087
Chemical Name or Material (+)-3,4,4a,5,6,10b-Hexahydro-2H-naphtho[1,2-b][1,4]oxazin-9-ol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.