missing translation for 'onlineSavingsMsg'
Learn More

3',4',7,8-Tetrahydroxyflavone, TRC

Product Code. 30453685
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453685 10 mg 10mg
30449031 100 mg 100mg
30452399 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453685 Supplier Toronto Research Chemicals Supplier No. TRCT79575810MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3440-24-2
Molecular Formula C15 H10 O6
Molecular Weight (g/mol) 286.24
IUPAC Name 2-(3,4-dihydroxyphenyl)-7,8-dihydroxy-chromen-4-one
SMILES Oc1ccc(cc1O)C2=CC(=O)c3ccc(O)c(O)c3O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H10O6/c16-9-3-1-7(5-12(9)19)13-6-11(18)8-2-4-10(17)14(20)15(8)21-13/h1-6,16-17,19-20H
Formula Weight 286.0477
Chemical Name or Material 3',4',7,8-Tetrahydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.