missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorosulfonylphenyl)propionic acid, 99%

Product Code. 11355369
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355369 1 g 1g
11365369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355369 Supplier Thermo Scientific Alfa Aesar Supplier No. H50384.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 63545-54-0 | C9H9ClO4S | 248.68 g/mol

3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63545-54-0
Molecular Formula C9H9ClO4S
Molecular Weight (g/mol) 248.68
MDL Number MFCD02179510
InChI Key INJMPXHPNFJMLG-UHFFFAOYSA-N
Synonym 3-4-chlorosulfonyl phenyl propanoic acid, benzenepropanoicacid, 4-chlorosulfonyl, 3-4-chlorosulphonyl phenyl propanoic acid, 3-4-chlorosulfonylphenyl propionic acid, 3-4-chlorosulfonylphenyl propanoic acid, p-chlorosylphonyldihydrocinnamic acid, 4-chlorosulfonyl benzenepropanoic acid, 4-chlorosulphonyl dihydrocinnamic acid, p-chlorosulphonyldihydrocinnamic acid
PubChem CID 2757915
IUPAC Name 3-[4-(chlorosulfonyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 248.69
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorosulfonylphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.