missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene Dihydrochloride, TRC

Product Code. 30350926
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350926 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350926 Supplier Toronto Research Chemicals Supplier No. TRCD4168502.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Diaminothiophene, Dihydrochloride
CAS 90069-81-1
Molecular Formula C4 H6 N2 S . 2 Cl H
Quantity 2.5 g
InChI Formula InChI=1S/C4H6N2S.2ClH/c5-3-1-7-2-4(3)6;;/h1-2H,5-6H2;2*1H
SMILES Cl.Cl.Nc1cscc1N
IUPAC Name thiophene-3,4-diamine;dihydrochloride
Molecular Weight (g/mol) 187.09
Formula Weight 185.9785
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.