Learn More
3,4-Dichloro-1,2,5-thiadiazole, TRC
Product Code. 30375382
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375382 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30375382 Supplier Toronto Research Chemicals Supplier No. TRCD435985100G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole
CAS 5728-20-1
Molecular Formula C2Cl2N2S
Quantity 100 g
Synonym 1,2,5-Thiadiazole, 3,4-dichloro-, 3,4-Dichloro-1,2,5-thiadiazole, 4,5-Dichloro-2,1,3-thiadiazole
InChI Formula InChI=1S/C2Cl2N2S/c3-1-2(4)6-7-5-1
SMILES Clc1nsnc1Cl
IUPAC Name 3,4-dichloro-1,2,5-thiadiazole
Molecular Weight (g/mol) 155.01
Formula Weight 153.9159