Learn More
3,4-Dichlorothiophenol, 97%
Product Code. 11338575
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338575 1 g 1g
11348575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338575 Supplier Thermo Scientific Alfa Aesar Supplier No. L12031.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
CAS: 5858-17-3 | C6H4Cl2S | 179.058 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5858-17-3
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00004840
InChI Key HNJZDPKMMZXSKT-UHFFFAOYSA-N
Synonym 3,4-dichlorothiophenol, 3,4-dichloro thiophenol, benzenethiol, 3,4-dichloro, 3,4-dichlorobenzene-1-thiol, pubchem6803, 3,4dichloro-thiophenol, acmc-1aset, 3,4-dichloro-thiophenol, 3 ,4-dichlorobenzenethiol, 3,4-dichlorophenylmercaptan
PubChem CID 79964
IUPAC Name 3,4-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1S)Cl)Cl
Specifications
Density 1.407
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.623
Quantity 1 g
Beilstein 2042869
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorothiophenol