missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™

Product Code. 11316589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11316589 5 g 5g
11306589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316589 Supplier Thermo Scientific Alfa Aesar Supplier No. H26186.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875664-54-3
Molecular Formula C6H7ClF2N2
Molecular Weight (g/mol) 180.58
MDL Number MFCD03094170
InChI Key QTEJTSFVIILHJJ-UHFFFAOYSA-N
Synonym 3,4-difluorophenylhydrazine hydrochloride, 3,4-difluorophenyl hydrazine hydrochloride, 3,4-difluorophenylhydrazinehydrochloride, 3,4-difluorophenylhydrazine hydrogen chloride, 3,4-difluorophenylhydrazine hcl, n'-3,4-difluoro-phenyl-hydrazinium, chloride, 3,4-difluorophenyl hydrazine hydrochloride 1:x, 3,4-difluoro-phenyl-hydrazine hydrochloride, hydrazine, 3,4-difluorophenyl-, hydrochloride
PubChem CID 24721344
IUPAC Name (3,4-difluorophenyl)hydrazine;hydrochloride
SMILES Cl.NNC1=CC(F)=C(F)C=C1

Specifications

Melting Point ∼230°C (decomposition)
Quantity 5 g
Formula Weight 180.59
Percent Purity 98%
Chemical Name or Material 3,4-Difluorophenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.