missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzyl Alcohol, TRC

Product Code. 30132626
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132626 100 mg 100mg
30137330 500 mg 500mg
30006268 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132626 Supplier Toronto Research Chemicals Supplier No. TRCD493205100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3897-89-0
Molecular Formula C7 H8 O3
Molecular Weight (g/mol) 140.14
IUPAC Name 5-(hydroxymethyl)benzene-1,3-diol
SMILES OCc1cc(O)cc(O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H8O3/c8-4-5-1-6(9)3-7(10)2-5/h1-3,8-10H,4H2
Formula Weight 140.0473
Chemical Name or Material 3,4-Dihydroxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.