missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Product Code. 11389015
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389015 100 mg 100mg
11379015 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389015 Supplier Thermo Scientific Alfa Aesar Supplier No. L14161.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4143-64-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00017600
InChI Key SRNPMQHYWVKBAV-UHFFFAOYSA-N
Synonym 3',4'-dihydroxyflavone, 2-3,4-dihydroxyphenyl-4h-chromen-4-one, 2-3,4-dihydroxyphenyl chromen-4-one, unii-koh101s66v, 3',4'-dihydroxy flavone, 3,4-dihydroxyflavone, 4h-1-benzopyran-4-one, 2-3,4-dihydroxyphenyl, dihydroxyflavone, 3',4', 4hlm, pubchem9842
PubChem CID 145726
IUPAC Name 2-(3,4-dihydroxyphenyl)chromen-4-one
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C3=CC(=C(C=C3)O)O

Specifications

Melting Point 249°C to 252°C
Quantity 100 mg
Beilstein 212540
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 3',4'-Dihydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.