missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzylamine, 97%

Product Code. 11465997
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465997 5 g 5g
11475997 25 g 25g
11485997 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465997 Supplier Thermo Scientific Alfa Aesar Supplier No. B25033.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5763-61-1 | C9H13NO2 | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5763-61-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00008116
InChI Key DIVNUTGTTIRPQA-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
PubChem CID 79832
IUPAC Name (3,4-dimethoxyphenyl)methanamine
SMILES COC1=CC=C(CN)C=C1OC

Specifications

Density 1.109
Boiling Point 281°C to 284°C
Flash Point >110°C (230°F)
Refractive Index 1.556
Quantity 5 g
UN Number UN2735
Beilstein 511575
Sensitivity Air Sensitive
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.