Learn More
(3,4-Dimethoxyphenyl)acetonitrile(Homoveratronitrile), TRC
Product Code. 30403252
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403252 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30403252 Supplier Toronto Research Chemicals Supplier No. TRCH669530250G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,4-Dimethoxyphenylacetonitrile
CAS 93-17-4
Molecular Formula C10 H11 N O2
Quantity 250 g
Synonym 3,4-Dimethoxyphenylacetonitrile, Benzeneacetonitrile, 3,4-dimethoxy-, Acetonitrile, (3,4-dimethoxyphenyl)- (6CI,7CI,8CI), 3,4-Dimethoxybenzeneacetonitrile, (3,4-Dimethoxyphenyl)acetonitrile, 2-(3,4-Dimethoxyphenyl)acetonitrile, 3,4-Dimethoxybenzyl cyanide, 3,4-Methyleneoxyphenyl acetonitrile, Homoveratronitrile, NSC 6324, Veratryl cyanide
InChI Formula InChI=1S/C10H11NO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5H2,1-2H3
SMILES COc1ccc(CC#N)cc1OC
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
Molecular Weight (g/mol) 177.2
Formula Weight 177.079