missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxytoluene, 98%

Product Code. 11483007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483007 5 g 5g
11493007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483007 Supplier Thermo Scientific Alfa Aesar Supplier No. A15558.06
  • ONLINE EXCLUSIVE PRICE
    49.20€
    44.65€ / 5g
    Save 4.55€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 494-99-5 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-99-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00016651
InChI Key GYPMBQZAVBFUIZ-UHFFFAOYSA-N
Synonym 3,4-dimethoxytoluene, homoveratrole, 4-methylveratrole, benzene, 1,2-dimethoxy-4-methyl, 4-methylveratrol, 4-methyl-1,2-dimethoxybenzene, 1,2-dimethoxy-4-methyl-benzene, unii-349x0g2ssf, homoveratrol
PubChem CID 68126
IUPAC Name 1,2-dimethoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)OC

Specifications

Melting Point 25°C to 28°C
Density 1.059
Boiling Point 220°C
Flash Point 84°C (183°F)
Odor Aromatic
Refractive Index 1.528
Quantity 5 g
Beilstein 2045328
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxytoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.