Learn More
3,4-Dimethylthiophene, 97%
Product Code. 15224268 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15224268 1 g 1g
15234268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15224268 Supplier Thermo Scientific Acros Supplier No. 450950010
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
CAS: 632-15-5 | C6H8S | 112.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 632-15-5
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD01764731
InChI Key GPSFYJDZKSRMKZ-UHFFFAOYSA-N
PubChem CID 79089
IUPAC Name 3,4-dimethylthiophene
SMILES CC1=CSC=C1C
Specifications
Boiling Point 144.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym thiophene, 3,4-dimethyl, 3,4-dimethyl-thiophene, 3,4-dimethyl thiophene, thiophene,3,4-dimethyl
Formula Weight 112.19
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylthiophene
compliance-icons compliance-icons
Product Identifier
  • 3,4-Dimethylthiophene, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.