missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxy-3,5-dimethoxyphenyl)propanoic Acid, TRC

Product Code. 30403050
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403050 100 mg 100mg
30400977 50 mg 50mg
30388808 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403050 Supplier Toronto Research Chemicals Supplier No. TRCH512475100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14897-78-0
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.23
IUPAC Name 3-(4-hydroxy-3,5-dimethoxyphenyl)propanoic acid
SMILES COc1cc(CCC(=O)O)cc(OC)c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H14O5/c1-15-8-5-7(3-4-10(12)13)6-9(16-2)11(8)14/h5-6,14H,3-4H2,1-2H3,(H,12,13)
Formula Weight 226.0841
Chemical Name or Material 3-(4-Hydroxy-3,5-dimethoxyphenyl)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.