missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Isobutylphenyl)propanoic Acid, TRC

Product Code. 30408557
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408557 1 g
30390853 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408557 Supplier Toronto Research Chemicals Supplier No. TRCI7803351G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65322-85-2
Molecular Formula C13 H18 O2
Molecular Weight (g/mol) 206.28
Synonym 3-[4-(2-Methylpropyl)phenyl]propanoic Acid, Ibuprofen Imp. F (EP)
IUPAC Name 3-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)Cc1ccc(CCC(=O)O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H18O2/c1-10(2)9-12-5-3-11(4-6-12)7-8-13(14)15/h3-6,10H,7-9H2,1-2H3,(H,14,15)
Formula Weight 206.1307
Percent Purity >95%
Chemical Name or Material 3-[4-(2-methylpropyl)phenyl]propanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.