missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Methoxyphenyl)-1H-pyrazole, 98%

Product Code. 15466287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466287 5 g 5g
15456287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466287 Supplier Alfa Aesar™ Supplier No. H33479.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 27069-17-6
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.20
MDL Number MFCD00112008
InChI Key QUSABYOAMXPMQH-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1h-pyrazole, 3-4-methoxyphenyl pyrazole, 5-4-methoxyphenyl-1h-pyrazole, chembl39210, 3-4-methoxy-phenyl-1h-pyrazole, 1h-pyrazole, 3-4-methoxyphenyl, 4-methoxy-1-pyrazol-3-ylbenzene, 3-4-methoxyphenyl-2h-pyrazole, maybridge1_007801, 3-4-methoxyphenyl-pyrazole
PubChem CID 599984
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazole
SMILES COC1=CC=C(C=C1)C1=CC=NN1

Specifications

Melting Point 126°C to 129°C
Quantity 5 g
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxyphenyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.