missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)pyrazole, 98%, Thermo Scientific™

Product Code. 10096712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10096712 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10096712 Supplier Thermo Scientific Acros Supplier No. 387080010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 27069-17-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 127°C to 130°C
Color White to Yellow
Boiling Point 185°C (0.6 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H10N2O
MDL Number MFCD00112008
Quantity 1 g
Synonym 3-4-methoxyphenyl-1h-pyrazole, 3-4-methoxyphenyl pyrazole, 5-4-methoxyphenyl-1h-pyrazole, chembl39210, 3-4-methoxy-phenyl-1h-pyrazole, 1h-pyrazole, 3-4-methoxyphenyl, 4-methoxy-1-pyrazol-3-ylbenzene, 3-4-methoxyphenyl-2h-pyrazole, maybridge1_007801, 3-4-methoxyphenyl-pyrazole
Solubility Information Solubility in water: insoluble
InChI Key QUSABYOAMXPMQH-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C1=CC=NN1
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazole
Molecular Weight (g/mol) 174.20
PubChem CID 599984
Formula Weight 174.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Methoxyphenyl)pyrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.