missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)nitrobenzene, +98%, ACROS Organics™

Product Code. 10002600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10002600 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10002600 Supplier Thermo Scientific Acros Supplier No. 188520100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 328-75-6
CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C8H3F6NO2
Linear Formula (CF3)2C6H3NO2
Refractive Index 1.4260 to 1.4280 (20°C, 589nm)
MDL Number MFCD00000384
Quantity 10 g
Beilstein 05, III, 841
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol) 259.11
PubChem CID 67603
Formula Weight 259.1
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)nitrobenzene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Bis(trifluoromethyl)nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.