missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzoic Acid, TRC

Product Code. 30374305
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374305 500 mg
30381052 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374305 Supplier Toronto Research Chemicals Supplier No. TRCD493738500MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421-49-4
Molecular Formula C15 H22 O3
Molecular Weight (g/mol) 250.33
Synonym Benzoic acid, 3,5-di-tert-butyl-4-hydroxy- (6CI,8CI), 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzoic acid, 2,6-Di-tert-butyl-4-carboxyphenol, 3,5-Di-tert-butyl-4-hydroxybenzoic acid, 3,5-Ditert-butyl-4-hydroxybenzoic acid, 4-Carboxy-2,6-di-tert-butylphenol, 4-Hydroxy-3,5-di-tert-butylbenzoic acid, F 2, F 2 (antioxidant), 4-Carboxy-2,6-di-tert-butylphenol, 4-Hydroxy-3,5-di-tert-butylbenzoic Acid, F 2F 2 (Antioxidant), 3,5-Ditert-butyl-4-hydroxybenzoic Acid, 3,5-Bis(1,1-Dimethylethyl)-4-hydroxybenzoic Acid, 2,6-Di-tert-butyl-4-carboxyphenol, Benzoic acid, 3,5-di-tert-butyl-4-hydroxy- (6CI,8CI)
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzoic acid
SMILES CC(C)(C)c1cc(cc(c1O)C(C)(C)C)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H22O3/c1-14(2,3)10-7-9(13(17)18)8-11(12(10)16)15(4,5)6/h7-8,16H,1-6H3,(H,17,18)
Formula Weight 250.1569
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.